• USA Home
  • 55390 - α-Hydroxyisobutyric acid

EMAIL THIS PAGE TO A FRIEND
55390 Sigma-Aldrich

α-Hydroxyisobutyric acid

purum, ≥98.0% (HPLC)

Synonym: 2-Hydroxy-2-methylpropionic acid, 2-Methyllactic acid, alpha-Hydroxyisobutyric acid

  • CAS Number 594-61-6

  • Linear Formula (CH3)2C(OH)COOH

  • Molecular Weight 104.10

  •  Beilstein Registry Number 1744739

  •  EC Number 209-848-8

  •  MDL number MFCD00004459

  •  PubChem Substance ID 329758101

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   BWLBGMIXKSTLSX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
bp   84 °C/1.5 mmHg(lit.)
mp   76-80 °C(lit.)
  76-80 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?