55460 Sigma-Aldrich

Hydroxylamine hydrochloride

puriss. p.a., ACS reagent, ≥99.0% (RT)

DOWNLOAD MSDS (PDF)

Synonym: Hydroxylammonium chloride

  • CAS Number 5470-11-1

  • Linear Formula NH2OH · HCl

  • Molecular Weight 69.49

  •  Beilstein Registry Number 3539763

  •  EC Number 226-798-2

  •  MDL number MFCD00051089

  •  PubChem Substance ID 24879418

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Bioactive Small Molecules, Cell Biology,
Quality Level   PREMIUM
grade   ACS reagent
  puriss. p.a.
assay   ≥99.0% (RT)
ign. residue   ≤0.05% (as SO4)
pH   2.5-3.5 (25 °C, 50 mg/mL in H2O)
mp   155-157 °C (dec.)(lit.)
density   1.67 g/mL at 25 °C(lit.)
anion traces   sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Hydroxylamine hydrochloride

99.9999% trace metals basis

Hydroxylamine hydrochloride

ACS reagent, 98.0%

Hydroxylamine hydrochloride

ReagentPlus®, 99%

Hydroxylamine hydrochloride

for AAS, ≥99.0%

Hydroxylamine hydrochloride

purum p.a., ≥98.0% (RT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?