• USA Home
  • 55469 - Hydroxylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
55469 Sigma-Aldrich

Hydroxylamine hydrochloride

purum p.a., ≥98.0% (RT)

Synonym: Hydroxylammonium chloride

  • CAS Number 5470-11-1

  • Linear Formula NH2OH · HCl

  • Molecular Weight 69.49

  •  Beilstein Registry Number 3539763

  •  EC Number 226-798-2

  •  MDL number MFCD00051089

Purchase

Properties

InChI Key   WTDHULULXKLSOZ-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥98.0% (RT)
mp   155-157 °C (dec.)(lit.)
density   1.67 g/mL at 25 °C(lit.)
anion traces   sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Description

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Signal word 
Warning
Precautionary statements 
RIDADR 
UN2923 - class 8 - PG 3 - EHS - acidic - Corrosive solids, toxic, n.o.s., HI: all
WGK Germany 
3
RTECS 
NC3675000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?