EMAIL THIS PAGE TO A FRIEND
55593 Sigma-Aldrich

Aluminum Ionophore I

Selectophore, function tested

Synonym: 2′,3,4′,5,7-Pentahydroxyflavone, Morin

  • CAS Number 480-16-0

  • Empirical Formula (Hill Notation) C15H10O7

  • Molecular Weight 302.24

  •  Colour Index Number 75660

  •  EC Number 207-542-9

  •  MDL number MFCD00006826

  •  PubChem Substance ID 329758139

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   YXOLAZRVSSWPPT-UHFFFAOYSA-N
quality   function tested

Description

Application

Application as aluminum(III) selective potentiometric sensor

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?