• USA Home
  • 55930 - 3-Hydroxy-2-naphthoic acid

EMAIL THIS PAGE TO A FRIEND
55930 Sigma-Aldrich

3-Hydroxy-2-naphthoic acid

technical, ≥95% (T)

Synonym: β-Hydroxynaphthoic acid

  • CAS Number 92-70-6

  • Linear Formula HOC10H6CO2H

  • Molecular Weight 188.18

  •  Beilstein Registry Number 744100

  •  EC Number 202-180-8

  •  MDL number MFCD00004103

  •  PubChem Substance ID 329758105

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   ALKYHXVLJMQRLQ-UHFFFAOYSA-N
grade   technical
assay   ≥95% (T)
mp   218-221 °C(lit.)
  219-222 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
QL1755000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?