EMAIL THIS PAGE TO A FRIEND
56191 Sigma-Aldrich

N-Hydroxyphthalimide

purum, ≥98.0% (T)

  • CAS Number 524-38-9

  • Empirical Formula (Hill Notation) C8H5NO3

  • Molecular Weight 163.13

  •  Beilstein Registry Number 131208

  •  EC Number 208-358-1

  •  MDL number MFCD00005891

  •  PubChem Substance ID 24879985

Purchase

Properties

Related Categories Active Esters/Additives, Chemical Synthesis, Coupling, Synthetic Reagents
InChI Key   CFMZSMGAMPBRBE-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   233 °C (dec.)(lit.)
  ~235 °C (dec.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TI5200000

Documents

Certificate of Analysis

Protocols & Articles

Articles

Coupling Reagents

In principle, the seemingly simple formation of a peptide bond can be accomplished using all the procedures available in organic chemistry for the synthesis of carboxylic acid amides. However, due to...
Matthias Junkers
Aldrich ChemFiles 2007, 7.2, 3.
Keywords: Acylations, Catalysis, Condensations, Dehydration reaction, Peptide synthesis, Racemizations, Solid phase peptide synthesis

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?