EMAIL THIS PAGE TO A FRIEND
56220 Sigma-Aldrich

4-Hydroxypiperidine

purum, ≥98.0% (NT)

Synonym: 4-Piperidinol

  • CAS Number 5382-16-1

  • Empirical Formula (Hill Notation) C5H11NO

  • Molecular Weight 101.15

  •  Beilstein Registry Number 102738

  •  EC Number 226-373-1

  •  MDL number MFCD00005999

  •  PubChem Substance ID 329758189

Purchase

Properties

Related Categories Building Blocks, C5 to C7, Chemical Synthesis, Heterocyclic Building Blocks, Piperidines More...
InChI Key   HDOWRFHMPULYOA-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (NT)
bp   108-114 °C/10 mmHg(lit.)
mp   86-90 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
226.4 °F
Flash Point(C) 
108 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?