• USA Home
  • 56270 - 3-Hydroxypropionitrile

EMAIL THIS PAGE TO A FRIEND
56270 Sigma-Aldrich

3-Hydroxypropionitrile

puriss., ≥99.0% (GC)

Synonym: 2-Cyanoethanol, Ethylene cyanohydrin, Hydracrylonitrile

  • CAS Number 109-78-4

  • Linear Formula HOCH2CH2CN

  • Molecular Weight 71.08

  •  Beilstein Registry Number 635773

  •  MDL number MFCD00002826

  •  PubChem Substance ID 329758196

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
vapor density   2.5 (vs air)
vapor pressure   <0.1 mmHg ( 20 °C)
InChI Key   WSGYTJNNHPZFKR-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
autoignition temp.   922 °F
expl. lim.   12.1 %
refractive index   n20/D 1.425(lit.)
  n20/D 1.426
bp   228 °C(lit.)
mp   −46 °C(lit.)
solubility   H2O: soluble0.5 g/5mL, clear, colorless
density   1.04 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MU5250000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?