• USA Home
  • 56296 - (−)-Δ9-Tetrahydrocannabinol solution

EMAIL THIS PAGE TO A FRIEND
56296 Sigma-Aldrich

(−)-Δ9-Tetrahydrocannabinol solution

~1 mg/mL in ethanol, analytical standard, for drug analysis

Synonym: Δ1-Tetrahydrocannabinol, Δ9-THC

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Cannabinoids Standards, Cannabiol,
grade   analytical standard, for drug analysis
InChI Key   CYQFCXCEBYINGO-IAGOWNOFSA-N
assay   ≥97% (HPLC)
shelf life   limited shelf life, expiry date on the label
drug control   USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)
concentration   ~1 mg/mL in ethanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   corresponds for H-NMR
format   single component solution
storage temp.   2-8°C

Description

Packaging

ampoule contains 1.2mL solution

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol, solution
WGK Germany 
2
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Active Cannabinoid Compounds in Gummy Bear Candy on Ascentis® Express Biphenyl with UV Detection after Extraction

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Active Cannabinoid Compounds in Gummy Bear Candy on Ascentis® Express Biphenyl with UV Detection after Extraction
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Cannabinoids using Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Cannabinoids using Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Separation, Solvents

UHPLC Analysis of Cannabinoids on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Cannabinoids on Ascentis® Express C18
Keywords: Chromatography, Clinical, Diseases, Forensic, Gas chromatography, High performance liquid chromatography, Mass spectrometry, Pharmaceutical, Separation

UHPLC/MS Analysis of Cannabinoids on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents UHPLC/MS Analysis of Cannabinoids on Ascentis® Express C18
Keywords: Clinical, Diseases, Forensic, Gas chromatography, High performance liquid chromatography, Mass spectrometry, Pharmaceutical, Separation, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules
08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?