• USA Home
  • 56510 - (±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
56510 Sigma-Aldrich

(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

purum, ≥98.0% (HPLC)

Synonym: Trolox

  • CAS Number 53188-07-1

  • Empirical Formula (Hill Notation) C14H18O4

  • Molecular Weight 250.29

  •  Beilstein Registry Number 1384051

  •  EC Number 258-422-8

  •  MDL number MFCD00006846

  •  PubChem Substance ID 24879938

Purchase

Properties

Related Categories Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   GLEVLJDDWXEYCO-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   187-189 °C(lit.)
  187-189 °C
Gene Information   rat ... Akr1b1(24192)

Description

Other Notes

Cytoprotective against oxyradicals

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DJ2273000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

238813 (±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?