• USA Home
  • 56610 - Dopamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
56610 Sigma-Aldrich

Dopamine hydrochloride

puriss., ≥98.5% (AT)

Synonym: 2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, 3,4-Dihydroxyphenethylamine hydrochloride, 3-Hydroxytyramine hydrochloride, 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride

  • CAS Number 62-31-7

  • Linear Formula (HO)2C6H3CH2CH2NH2·HCl

  • Molecular Weight 189.64

  •  Beilstein Registry Number 3656720

  •  EC Number 200-527-8

  •  MDL number MFCD00012898

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   CTENFNNZBMHDDG-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (AT)
form   powder
ign. residue   ≤0.05%
mp   ~245 °C (dec.)
  248-250 °C(lit.)
solubility   H2O: soluble1 g/10 mL at 20 °C, clear, colorless

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
UX1092000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

H8502 Dopamine hydrochloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?