EMAIL THIS PAGE TO A FRIEND
56718 Sigma-Aldrich

Iberiotoxin

from Buthus tamulus, ≥85% (HPLC)

  • CAS Number 129203-60-7

  • Empirical Formula (Hill Notation) C179H274N50O55S7

  • Molecular Weight 4230.85

  •  MDL number MFCD00131365

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Other More...
biological source   from Buthus tamulus
InChI Key   VDNVVLOBNHIMQA-UHFFFAOYSA-N
assay   ≥85% (HPLC)
solubility   H2O: soluble1 mg/mL, clear, colorless
storage temp.   −20°C

Description

Amino Acid Sequence

pGlu-Phe-Thr-Asp-Val-Asp-Cys-Ser-Val-Ser-Lys-Glu-Cys-Trp-Ser-Val-Cys-Lys-Asp-Leu-Phe-Gly-Val-Asp-Arg-Gly-Lys-Cys-Met-Gly-Lys-Lys-Cys-Arg-Cys-Tyr-Gln [Disulfide Bridges: 7−28; 13−33; 17−35]

Biochem/physiol Actions

Selective and reversible inhibitor of high-conductance calcium-activated potassium channel.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3462 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?