• USA Home
  • 56802 - 5H-Dibenz[b,f]azepine

EMAIL THIS PAGE TO A FRIEND
56802 Sigma-Aldrich

5H-Dibenz[b,f]azepine

purum, ≥97.0% (NT)

Synonym: Iminostilbene

  • CAS Number 256-96-2

  • Empirical Formula (Hill Notation) C14H11N

  • Molecular Weight 193.24

  •  Beilstein Registry Number 1343358

  •  EC Number 205-970-0

  •  MDL number MFCD00005071

  •  PubChem Substance ID 329758239

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, N-Containing, Other Heterocyclic Building Blocks More...
InChI Key   LCGTWRLJTMHIQZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (NT)
form   powder
color   yellow to orange
mp   196-199 °C(lit.)
  196-199 °C
solubility   dioxane: 50 mg/mL clear

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

143650 5H-Dibenz[b,f]azepine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?