EMAIL THIS PAGE TO A FRIEND
57182 Sigma-Aldrich

Indo 1-AM

~95% (HPLC)

Synonym: 4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester

Purchase

Properties

Related Categories Biochemicals and Reagents, Calcium Indicators, Chelators and Ion Probes, Chelators and Ion Probes for Flow Cytometry, Chelators and Ion Probes for Microscopy,
InChI Key   CAWBRCOBJNWRLK-UHFFFAOYSA-N
assay   ~95% (HPLC)
solubility   acetonitrile: soluble10 mg/mL, clear
fluorescence   λex 330 nm; λem 398 nm in 0.1 M Tris pH 8.0, 10 mM Ca2+ (after cleavage by esterase)
storage temp.   2-8°C

Description

Other Notes

Cell-permeant derivative of Indo 1; Quant. assessment of intracellular free calcium concns. using ratiometric fluorescence technique

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I3261 Indo 1-AM, ~95% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?