EMAIL THIS PAGE TO A FRIEND
57190 Sigma-Aldrich

Indole

puriss., ≥98.5% (GC)

Synonym: 1H-Benzo[b]pyrrole

  • CAS Number 120-72-9

  • Empirical Formula (Hill Notation) C8H7N

  • Molecular Weight 117.15

  •  Beilstein Registry Number 107693

  •  EC Number 204-420-7

  •  MDL number MFCD00005607

  •  PubChem Substance ID 329758278

Purchase

Properties

Related Categories Building Blocks, C7 to C9, Cell Biology, Chemical Synthesis, Citrus aurantium (Seville orange),
InChI Key   SIKJAQJRHWYJAI-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (GC)
bp   253-254 °C(lit.)
mp   50-53 °C
  51-54 °C(lit.)
solubility   methanol: soluble0.1 g/mL, clear

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
1
RTECS 
NL2450000
Flash Point(F) 
249.8 °F
Flash Point(C) 
121 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?