• USA Home
  • 57210 - Indole-3-carboxaldehyde

EMAIL THIS PAGE TO A FRIEND
57210 Sigma-Aldrich

Indole-3-carboxaldehyde

purum, ≥98.0% (T)

Synonym: β-Indolylaldehyde, 3-Indolylformaldehyde, 3-Formylindole, Indole-3-carbaldehyde, NSC 10118

  • CAS Number 487-89-8

  • Empirical Formula (Hill Notation) C9H7NO

  • Molecular Weight 145.16

  •  Beilstein Registry Number 114117

  •  EC Number 207-665-8

  •  MDL number MFCD00005622

  •  PubChem Substance ID 329758281

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C7 to C9, C9, Carbonyl Compounds,
InChI Key   OLNJUISKUQQNIM-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   193-198 °C(lit.)
  193-198 °C

Description

Application

Reactant for preparation of:
• Analgesic agents
• Hypoglycemic agents
• Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
• Antibacterial and antifungal agents
• Antiamoebic and cytotoxic agents
• Inhibitors of the Dengue virus protease with antiviral activity in cell-culture
• Curcumin analogues as possible anti-proliferative & anti-inflammatory agents
• Inhibitors of Bcl-2 family proteins
• Inhibitors of the C-terminal domain of RNA Polymerase II as antitumor agents
• Inhibitors of TNF-α and IL-6 with anti-tubercular activity

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NL5993600

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

129445 Indole-3-carboxaldehyde, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?