EMAIL THIS PAGE TO A FRIEND
57240 Sigma-Aldrich

Indoline

purum, ≥98.0% (GC)

Synonym: 1-Azaindan, 2,3-Dihydroindole

  • CAS Number 496-15-1

  • Empirical Formula (Hill Notation) C8H9N

  • Molecular Weight 119.16

  •  Beilstein Registry Number 111915

  •  EC Number 207-816-8

  •  MDL number MFCD00005705

  •  PubChem Substance ID 329758285

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Indolines
InChI Key   LPAGFVYQRIESJQ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
form   liquid (clear)
color   clear dark brown
refractive index   n20/D 1.592(lit.)
  n20/D 1.592
bp   220-221 °C(lit.)
density   1.063 g/mL at 25 °C(lit.)

Description

Application

Reactant for preparation of:
• Inhibitors of NOD1-Induced Nuclear Factor-κB Activation
• Sphingosine-1-phosphate 4(S1P4) receptor antagonists
• Cytotoxic cell cycle inhibitors
• 2-Aminopyridines
• PET agent for imaging of protein kinase C (PKC)
• Sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes
• α4β2-Nicotinic acetylcholine receptor-selective partial agonists
• mGlu4 positive allosteric modulators
• Bacterial biofilm inhibitors
• Serotonin 5-HT6 receptor antagonists

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
RTECS 
NL6906300
Flash Point(F) 
199.4 °F
Flash Point(C) 
93 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?