EMAIL THIS PAGE TO A FRIEND
57260 Sigma-Aldrich

2-Oxindole

purum, ≥97.0% (HPLC)

Synonym: 2-Indolinone, Oxindole

  • CAS Number 59-48-3

  • Empirical Formula (Hill Notation) C8H7NO

  • Molecular Weight 133.15

  •  Beilstein Registry Number 114692

  •  EC Number 200-429-5

  •  MDL number MFCD00005711

  •  PubChem Substance ID 329758287

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Indolines
InChI Key   JYGFTBXVXVMTGB-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
bp   227 °C/73 mmHg(lit.)
mp   123-128 °C(lit.)
  123-128 °C
Gene Information   human ... PGR(5241)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NM2080500
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?