EMAIL THIS PAGE TO A FRIEND
57280 Sigma-Aldrich

3-Indoleacetonitrile

purum, ≥96.0% (GC)

Synonym: (3-Indolyl)acetonitrile, 3-(Cyanomethyl)indole, IAN, Indolylacetonitrile, NSC 523272

  • CAS Number 771-51-7

  • Empirical Formula (Hill Notation) C10H8N2

  • Molecular Weight 156.18

  •  Beilstein Registry Number 125488

  •  EC Number 212-232-1

  •  MDL number MFCD00005628

  •  PubChem Substance ID 24880661

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   DMCPFOBLJMLSNX-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
bp   157-160 °C/0.2 mmHg(lit.)
mp   33-36 °C(lit.)
  33-36 °C
storage temp.   2-8°C

Description

Other Notes

Plant growth activator. Promotes callus growth and shoot formation in tobacco callus

Same product — same quality — now Sigma-Aldrich brand

Application

Reactant for preparation of:
• Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
• Histone deacetylase inhibitors
• Potential kinase inhibitors
• Kv7/KCNQ potassium channel activators
• Kinesin-Specific MKLP-2 Inhibitor
• Pesticides
• Potential PET cancer imaging agents
• Agonists of the Farnesoid X Receptor (FXR) as atherosclerosis treatment
• Butyrylcholinesterase inhibitors
• Necroptosis inhibitors

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AM0700000
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C
Protocols & Articles

Articles

Indoles

Sigma-Aldrich is proud to offer a new series of ChemFiles—called Privileged Structures, to our Drug Discovery and Organic Synthesis customers. Each piece will highlight a specific motif, selected app...
Chemfiles Volume 4 Article 8
Keywords: Applications, Building blocks, Cardiovascular, Chemfiles, Company, Diabetes, Diseases, Drug discovery, Medicinal chemistry, Organic synthesis, Respiratory

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?