EMAIL THIS PAGE TO A FRIEND
57290 Sigma-Aldrich

3-Indoleacrylic acid

purum, ≥97.0% (HPLC)

Synonym: 3-(3-Indolyl)acrylic acid, IAA

  • CAS Number 1204-06-4

  • Empirical Formula (Hill Notation) C11H9NO2

  • Molecular Weight 187.19

  •  Beilstein Registry Number 6317

  •  EC Number 214-872-7

  •  MDL number MFCD00005633

  •  PubChem Substance ID 329758294

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   PLVPPLCLBIEYEA-AATRIKPKSA-N
grade   purum
assay   ≥97.0% (HPLC)
mp   ~190 °C (dec.)

Description

Other Notes

Tryptophan anti-metabolite, inhibits the conversion of indole to tryptophan by tryptophan synthetase in Sacchamomyces cerevisiae

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NL3680000

Documents

Certificate of Analysis

Protocols & Articles

Articles

Indoles

Sigma-Aldrich is proud to offer a new series of ChemFiles—called Privileged Structures, to our Drug Discovery and Organic Synthesis customers. Each piece will highlight a specific motif, selected app...
Chemfiles Volume 4 Article 8
Keywords: Applications, Building blocks, Cardiovascular, Chemfiles, Company, Diabetes, Diseases, Drug discovery, Medicinal chemistry, Organic synthesis, Respiratory

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?