• USA Home
  • 57300 - Indole-3-pyruvic acid

EMAIL THIS PAGE TO A FRIEND
57300 Sigma-Aldrich

Indole-3-pyruvic acid

≥98.0% (TLC)

Synonym: 3-(3-Indolyl)-2-oxopropanoic acid, 3-Indolylpyruvic acid

  • CAS Number 392-12-1

  • Empirical Formula (Hill Notation) C11H9NO3

  • Molecular Weight 203.19

  •  Beilstein Registry Number 172966

  •  EC Number 206-874-1

  •  MDL number MFCD00005640

  •  PubChem Substance ID 24880669

Purchase

Properties

InChI Key   RSTKLPZEZYGQPY-UHFFFAOYSA-N
assay   ≥98.0% (TLC)
ign. residue   ≤0.1%
mp   215 °C (dec.)(lit.)
  ~215 °C (dec.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NM1880000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

I7017 Indole-3-pyruvic acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?