EMAIL THIS PAGE TO A FRIEND
57333 Sigma-Aldrich

3-Indoleacetic acid

technical, ≥95% (T)

Synonym: Heteroauxin, IAA

  • CAS Number 87-51-4

  • Empirical Formula (Hill Notation) C10H9NO2

  • Molecular Weight 175.18

  •  Beilstein Registry Number 143358

  •  EC Number 201-748-2

  •  MDL number MFCD00005636

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   SEOVTRFCIGRIMH-UHFFFAOYSA-N
grade   technical
assay   ≥95% (T)
mp   165-169 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NL3150000

Documents

Certificate of Analysis

Protocols & Articles

Articles

Indoles

Sigma-Aldrich is proud to offer a new series of ChemFiles—called Privileged Structures, to our Drug Discovery and Organic Synthesis customers. Each piece will highlight a specific motif, selected app...
Chemfiles Volume 4 Article 8
Keywords: Applications, Building blocks, Cardiovascular, Chemfiles, Company, Diabetes, Diseases, Drug discovery, Medicinal chemistry, Organic synthesis, Respiratory

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?