EMAIL THIS PAGE TO A FRIEND
57412 Sigma-Aldrich

Indophenol

for microscopy

Synonym: Phenolindophenol

  • CAS Number 500-85-6

  • Empirical Formula (Hill Notation) C12H9NO2

  • Molecular Weight 199.21

  •  Beilstein Registry Number 2095656

  •  EC Number 207-913-5

  •  MDL number MFCD00001621

  •  PubChem Substance ID 329758307

Purchase

Properties

grade   for microscopy
InChI Key   RSAZYXZUJROYKR-UHFFFAOYSA-N
mp   >300 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
GU5778000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?