EMAIL THIS PAGE TO A FRIEND
57413 Sigma-Aldrich

Indomethacin

≥99.0% (TLC)

Synonym: 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-3-indoleacetic acid

  • CAS Number 53-86-1

  • Empirical Formula (Hill Notation) C19H16ClNO4

  • Molecular Weight 357.79

  •  Beilstein Registry Number 497341

  •  EC Number 200-186-5

  •  MDL number MFCD00057095

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics G-M, Biochemicals and Reagents, Inhibits an Enzyme,
InChI Key   CGIGDMFJXJATDK-UHFFFAOYSA-N
assay   ≥99.0% (TLC)
mp   155-162 °C
solubility   ethanol: soluble50 mg/mL, clear, yellow-green

Description

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that is relatively selective for COX-1.

Other Notes

Blocks prostaglandin biosynthesis.; Topography of prostaglandin H synthase; Cyclooxygenase inhibitor

This product has been replaced by I7378-SIGMA | Indomethacin ≥99% (TLC)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGI
WGK Germany 
3
RTECS 
NL3500000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

I7378 Indomethacin, ≥99% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?