• USA Home
  • 57462 - Infusion powder from bovine heart

EMAIL THIS PAGE TO A FRIEND
57462 Sigma-Aldrich

Infusion powder from bovine heart

for microbiology

Purchase

Properties

Related Categories Base Ingredients for Media, Microbiology, Protein Sources (Peptones & Extracts), Protein Sources A to Z
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
impurities   ≤12% ash
  ≥11% total nitrogen
  ≥3% α-amino nitrogen
loss   ≤10% loss on drying
pH   6-7.5 (25 °C)
solubility   H2O: soluble2%, clear, slightly yellow to brown

Description

Application

The inclusion of bovine heart infusion in the corresponding medium provides the necessary additional nutrients for the cultivation of fastidious microorganisms like Streptococci, Pneumococci, Meningcocci, Brucella species.

Other Notes

Sales restrictions may apply

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology's Basic Ingredient: Protein Sources (Peptones)

Peptones and extracts are excellent natural sources of amino acids, peptides and proteins in growth media. They are most often obtained by enzymatic digestion or acid hydrolysis of natural products, ...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?