EMAIL THIS PAGE TO A FRIEND
57575 Sigma-Aldrich

myo-Inositol

purum, ≥98.0% (HPLC)

Synonym: 1,2,3,4,5,6-Hexahydroxycyclohexane, i-Inositol, meso-Inositol

  • CAS Number 87-89-8

  • Empirical Formula (Hill Notation) C6H12O6

  • Molecular Weight 180.16

  •  Beilstein Registry Number 1907329

  •  EC Number 201-781-2

  •  MDL number MFCD00077932

  •  PubChem Substance ID 329758320

Purchase

Properties

Related Categories Artemisia vulgaris, Carbohydrate Chemistry, Carbohydrate Derivatives, Carbohydrates, Cell Biology,
vapor density   6.2 (vs air)
InChI Key   CDAISMWEOUEBRE-GPIVLXJGSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   222-227 °C(lit.)
  222-227 °C
solubility   H2O: soluble0.1 g/mL, clear

Description

Biochem/physiol Actions

A component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers.

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?