EMAIL THIS PAGE TO A FRIEND
57610 Sigma-Aldrich

Inulin from chicory

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates H-L, Cell Biology,
InChI Key   TUULCWHRUVPVEB-UGWHTDGHSA-N
ign. residue   ≤0.5%
loss   ~10% loss on drying

Description

Application

Potential replacement for high fructose corn syrup due to its prebiotic function and similar structural characteristics to corn syrup.

Biochem/physiol Actions

A heterogeneous blend of fructose polymers that function as plant storage carbohydrates.

Other Notes

Raw material for the production of fructose syrups, a review

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I2255 Inulin from chicory

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?