• USA Home
  • 57614 - Inulin from dahlia tubers

EMAIL THIS PAGE TO A FRIEND
57614 Sigma-Aldrich

Inulin from dahlia tubers

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrate adjuvants, Carbohydrates, Carbohydrates A to Z, Carbohydrates H-L,
InChI Key   TUULCWHRUVPVEB-UGWHTDGHSA-N
optical activity   [α]20/D −37.0±2°, c = 6.5% in H2O
mol wt   Mr ~5000
ign. residue   ≤0.2%
loss   ~5% loss on drying

Description

Other Notes

Polysaccharide reserve of Compositae.; Inulin containing plants as alternative resources; Suitable for the production of fructose syrups, review; Potentials for non-food utilization of fructose and inulin, review

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I3754 Inulin from dahlia tubers

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?