EMAIL THIS PAGE TO A FRIEND
57673 Sigma-Aldrich

Iodoacetonitrile

purum, ≥96.0% (GC)

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   VODKOOOHHCAWFR-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
refractive index   n20/D 1.574(lit.)
  n20/D 1.576
bp   182-184 °C/720 mmHg(lit.)
density   2.307 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

I6903 Iodoacetonitrile, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?