• USA Home
  • 57674 - 5-(Iodoacetamido)fluorescein

EMAIL THIS PAGE TO A FRIEND
57674 Sigma-Aldrich

5-(Iodoacetamido)fluorescein

for fluorescence, ≥80.0% (HPLC)

Synonym: 5-IAF

  • CAS Number 63368-54-7

  • Empirical Formula (Hill Notation) C22H14INO6

  • Molecular Weight 515.25

  •  Beilstein Registry Number 6543244

Purchase

Description

Other Notes

Fluorescent thiol reagent.; Fluorescent measurement of desmin intermediate filament assembly; Label in flow cytofluorometric analysis

Same product — same quality — now Sigma-Aldrich brand

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

I9271 5-(Iodoacetamido)fluorescein, ≥90% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?