EMAIL THIS PAGE TO A FRIEND
57675 Sigma-Aldrich

2-Iodoethanol

purum, ≥99.0% (GC)

  • CAS Number 624-76-0

  • Linear Formula ICH2CH2OH

  • Molecular Weight 171.97

  •  Beilstein Registry Number 1098258

  •  EC Number 210-861-6

  •  MDL number MFCD00002830

  •  PubChem Substance ID 24880881

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
InChI Key   QSECPQCFCWVBKM-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
contains   copper turnings as stabilizer
refractive index   n20/D 1.572(lit.)
  n20/D 1.572
bp   85 °C/25 mmHg(lit.)
density   2.20 g/mL at 20 °C
  2.205 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
3
RTECS 
KL3820000
Flash Point(F) 
149 °F
Flash Point(C) 
65 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

176850 2-Iodoethanol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?