EMAIL THIS PAGE TO A FRIEND
57690 Sigma-Aldrich

3-Iodoaniline

purum, ≥97.0% (NT)

Purchase

Properties

InChI Key   FFCSRWGYGMRBGD-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (NT)
form   liquid (clear)
color   clear yellow
refractive index   n20/D 1.682(lit.)
bp   145-146 °C/15 mmHg(lit.)
mp   21-24 °C(lit.)
density   1.821 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
BY3750000
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I7209 3-Iodoaniline, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?