EMAIL THIS PAGE TO A FRIEND
57730 Sigma-Aldrich

2-Iodobenzoic acid

purum, ≥95.0% (GC)

  • CAS Number 88-67-5

  • Linear Formula IC6H4CO2H

  • Molecular Weight 248.02

  •  Beilstein Registry Number 1861406

  •  EC Number 201-850-7

  •  MDL number MFCD00002419

  •  PubChem Substance ID 329758349

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   CJNZAXGUTKBIHP-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
ign. residue   ≤0.5%
mp   160-162 °C(lit.)
  161-164 °C

Description

Other Notes

Starting material for the preparation of iodosobenzoate, a useful oxidant. Protecting group for alcohols, the esters are cleaved oxidatively

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DH2975000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I7675 2-Iodobenzoic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?