EMAIL THIS PAGE TO A FRIEND
57731 Sigma-Aldrich

3-Iodobenzoic acid

purum, ≥98.0% (HPLC)

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   KVBWBCRPWVKFQT-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
ign. residue   ≤0.05%
mp   185-187 °C(lit.)
  186-188 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

138584 3-Iodobenzoic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?