EMAIL THIS PAGE TO A FRIEND
57735 Sigma-Aldrich

4-Iodobenzoic acid

purum, ≥97.0% (HPLC)

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   GHICCUXQJBDNRN-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
ign. residue   ≤0.1%
mp   270-273 °C(lit.)
  270-273 °C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DH2976000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

206547 4-Iodobenzoic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?