• USA Home
  • 57745 - 2-Iodobenzoyl chloride

EMAIL THIS PAGE TO A FRIEND
57745 Sigma-Aldrich

2-Iodobenzoyl chloride

technical, ≥97.0% (AT)

  • CAS Number 609-67-6

  • Linear Formula IC6H4COCl

  • Molecular Weight 266.46

  •  Beilstein Registry Number 2042672

  •  EC Number 210-196-1

  •  MDL number MFCD00001040

  •  PubChem Substance ID 24880562

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   MVIVDSWUOGNODP-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (AT)
bp   105-106 °C/1 mmHg(lit.)
mp   27-31 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?