EMAIL THIS PAGE TO A FRIEND
57840 Sigma-Aldrich

Iodoacetic acid

≥98.0% (T)

  • CAS Number 64-69-7

  • Linear Formula ICH2CO2H

  • Molecular Weight 185.95

  •  Beilstein Registry Number 1739079

  •  EC Number 200-590-1

  •  MDL number MFCD00002685

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Organic Acids, Synthetic Reagents More...
InChI Key   JDNTWHVOXJZDSN-UHFFFAOYSA-N
assay   ≥98.0% (T)
color   faintly yellow
mp   77-79 °C(lit.)
  80-84 °C
storage temp.   2-8°C

Description

Application

Reagent for the modification of cysteine residues in proteins.

Other Notes

For the chemical modification of thiol groups in proteins (specific for cysteine at pH 8.5) ; Alkylates methionine

This product has been replaced by I4386-Sigma-Aldrich | Iodoacetic acid ≥98.0% (T)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2923 6.1(8) / PGI
WGK Germany 
1
RTECS 
AI3500000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?