EMAIL THIS PAGE TO A FRIEND
57850 Sigma-Aldrich

Ethyl iodoacetate

purum, ≥98.0% (GC)

Purchase

Properties

Related Categories Building Blocks, C2 to C5, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   MFFXVVHUKRKXCI-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.503(lit.)
  n20/D 1.506
bp   179-180 °C(lit.)
density   1.808 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2922 6.1(8) / PGII
WGK Germany 
3
RTECS 
AI3575000
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

242934 Ethyl iodoacetate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?