EMAIL THIS PAGE TO A FRIEND
57854 Sigma-Aldrich

Iodoacetic anhydride

technical, ≥90% (T)

  • CAS Number 54907-61-8

  • Linear Formula (ICH2CO)2O

  • Molecular Weight 353.88

  •  Beilstein Registry Number 1812140

  •  MDL number MFCD00001080

  •  PubChem Substance ID 24880933

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Anhydrides, Chemical Synthesis, Organic Building Blocks More...
InChI Key   RBNSZWOCWHGHMR-UHFFFAOYSA-N
grade   technical
assay   ≥90% (T)
mp   47-49 °C(lit.)
  47-49 °C
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Reagent used as linker for development of reagents used for differential protein quantitation via ion scanning

Reactant used for:
• Sythesis of N-iodoacetyl glycosylamine derivatives and converting amino precursors to IA derivatives
• Linking lysine residues to N-terminal α-amino groups of peptides
• Capping amines and yielding a thiol reactive iodo-derivative
• Iodoacetylation

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3261 8/PG 2
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

284262 Iodoacetic anhydride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?