EMAIL THIS PAGE TO A FRIEND
57870 Sigma-Aldrich

1-Iodoheptane

purum, ≥98.0% (GC)

Synonym: Heptyl iodide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   LMHCYRULPLGEEZ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.490(lit.)
  n20/D 1.490
bp   204 °C(lit.)
mp   −48 °C(lit.)
density   1.379 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

177857 1-Iodoheptane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?