EMAIL THIS PAGE TO A FRIEND
57890 Sigma-Aldrich

1-Iodohexane

purum, ≥97.0% (GC)

Synonym: Hexyl iodide

  • CAS Number 638-45-9

  • Linear Formula CH3(CH2)5I

  • Molecular Weight 212.07

  •  Beilstein Registry Number 505971

  •  EC Number 211-339-0

  •  MDL number MFCD00001102

  •  PubChem Substance ID 24880957

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   ANOOTOPTCJRUPK-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
contains   silver wool as stabilizer
refractive index   n20/D 1.492(lit.)
  n20/D 1.492
bp   179-180 °C(lit.)
density   1.437 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3082 - class 9 - PG 3 - DOT NA1993 - Environmentally hazardous substances, liquid, n.o.s. HI: all (not BR)
WGK Germany 
2
Flash Point(F) 
141.8 °F
Flash Point(C) 
61 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

238287 1-Iodohexane, contains copper as stabilizer, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?