• USA Home
  • 57895 - Iodide polymer supported

EMAIL THIS PAGE TO A FRIEND
57895 Sigma-Aldrich

Iodide polymer supported

extent of labeling: ~2.9 mmol/g I- loading

Synonym: Amberlite® IRA 900 I--form

Purchase

Properties

Related Categories Chemical Synthesis, Other Supported Reagents, Solid Supported Synthesis, Specialty Synthesis, Supported Reagents More...
extent of labeling   ~2.9 mmol/g I- loading
impurities   ≤2% water

Description

Other Notes

Polymer supported reagent for the preparation of alkyl iodides from other alkyl halides; Starting material for polymer supported chlorine

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?