EMAIL THIS PAGE TO A FRIEND
57910 Sigma-Aldrich

1-Iodonaphthalene

purum, ≥96.0% (GC)

Synonym: 1-Naphthyl iodide

  • CAS Number 90-14-2

  • Empirical Formula (Hill Notation) C10H7I

  • Molecular Weight 254.07

  •  Beilstein Registry Number 1906415

  •  EC Number 201-968-9

  •  MDL number MFCD00003876

  •  PubChem Substance ID 24880969

Purchase

Properties

Related Categories Aryl, Building Blocks, C9 to C12, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   NHPPIJMARIVBGU-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
form   liquid (clear)
color   clear very deep brown-yellow
refractive index   n20/D 1.701(lit.)
bp   163-165 °C/15 mmHg(lit.)
density   1.74 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?