Synonym:
2-
CAS Number 111-77-3
Linear Formula CH3OCH2CH2OCH2CH2OH
Molecular Weight 120.15
Beilstein Registry Number 1697812
EC Number 203-906-6
MDL number MFCD00002871
PubChem Substance ID 24880998
Popular Documents: Specification Sheet (PDF)
| grade | ReagentPlus® |
| vapor density | 4.14 (vs air) |
| vapor pressure | 0.2 mmHg ( 20 °C) |
| assay | ≥99.0% |
| autoignition temp. | 221 °C |
| expl. lim. | 1.38-22.7 % |
| refractive index |
n |
| bp | 194 °C(lit.) |
1, 4 L in poly bottle
Product of Dow Chemical.
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
CARBITOL is a trademark of The Dow Chemical Company or an affiliated company of Dow
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?