EMAIL THIS PAGE TO A FRIEND
57971 Sigma-Aldrich

1-Iodooctane

purum, ≥97.0% (GC)

Synonym: Octyl iodide

  • CAS Number 629-27-6

  • Linear Formula CH3(CH2)7I

  • Molecular Weight 240.13

  •  Beilstein Registry Number 1697479

  •  EC Number 211-084-5

  •  MDL number MFCD00001105

  •  PubChem Substance ID 24881007

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   UWLHSHAHTBJTBA-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
contains   silver wool as stabilizer
refractive index   n20/D 1.4878(lit.)
  n20/D 1.488
bp   225-226 °C(lit.)
mp   −46-−45 °C(lit.)
density   1.33 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
RTECS 
RG9700100
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?