EMAIL THIS PAGE TO A FRIEND
58010 Sigma-Aldrich

1-Iodopentane

purum, ≥98.0% (GC)

Synonym: n-Amyl iodide, Pentyl iodide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   BLXSFCHWMBESKV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
contains   copper as stabilizer
refractive index   n20/D 1.495(lit.)
  n20/D 1.495
bp   154-155 °C(lit.)
density   1.517 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
SA2975600
Flash Point(F) 
109.4 °F
Flash Point(C) 
43 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

241946 1-Iodopentane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?