EMAIL THIS PAGE TO A FRIEND
58020 Sigma-Aldrich

4-Iodophenol

purum, ≥98.0% (GC)

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   VSMDINRNYYEDRN-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   138 °C/5 mmHg(lit.)
mp   92-94 °C(lit.)
  92-94 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3
RTECS 
SL5600000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I10201 4-Iodophenol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?