• USA Home
  • 58032 - 4-Iodo-L-phenylalanine

EMAIL THIS PAGE TO A FRIEND
58032 Sigma-Aldrich

4-Iodo-L-phenylalanine

purum, ≥96.0% (HPLC), ~1 mol/mol water

Synonym: (S)-2-Amino-3-(4-iodophenyl)propanoic acid

  • CAS Number 24250-85-9

  • Empirical Formula (Hill Notation) C9H10INO2

  • Molecular Weight 291.09

  •  Beilstein Registry Number 4411317

  •  MDL number MFCD00002602

  •  PubChem Substance ID 24881029

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Phenylalanine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   PZNQZSRPDOEBMS-QMMMGPOBSA-N
grade   purum
assay   ≥96.0% (HPLC)
optical activity   [α]20/D −6.5±2°, c = 1% in acetic acid: water (4:1)
impurities   ~1 mol/mol water

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?