EMAIL THIS PAGE TO A FRIEND
58070 Sigma-Aldrich

N-Iodosuccinimide

purum, ≥97.0% (RT)

  • CAS Number 516-12-1

  • Empirical Formula (Hill Notation) C4H4INO2

  • Molecular Weight 224.98

  •  Beilstein Registry Number 113917

  •  EC Number 208-221-6

  •  MDL number MFCD00005512

  •  PubChem Substance ID 24881033

Purchase

Properties

Related Categories C-X Bond Formation (Halogen), Chemical Synthesis, Iodination, Synthetic Reagents
InChI Key   LQZMLBORDGWNPD-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (RT)
mp   ~200 °C (dec.)
  202-206 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Reagent for the oxidimetric titration of sulfur functions ; Reagent for oxidations ; Reagent for iodinations

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
WN2817000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

220051 N-Iodosuccinimide, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?