EMAIL THIS PAGE TO A FRIEND
58110 Sigma-Aldrich

4-Iodotoluene

purum, ≥98.0% (GC)

Synonym: 1-Iodo-4-methylbenzene

Purchase

Properties

Related Categories Aryl, Building Blocks, C7, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   UDHAWRUAECEBHC-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   211.5 °C(lit.)
mp   33-35 °C(lit.)
  33-35 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?